Learn More
Eurofins Discoverx ChemiScreen CaS Calcium Sensor Receptor Stable Cell Line, 2 vials. (HTS137C)
Catalog No. 502005957
Encompass
The ChemiScreen CaS Calcium Sensor Receptor Stable Cell Line is an engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. extracellular calcium) based activation of CASR GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses CASR alongside endogenous high levels of the promiscuous G protein, Gα15, coupling the receptor to the calcium signaling pathway. Upon activation of CASR, the resulting calcium activity can be fluorescently assayed. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, visit www.discoverx.com/targets/gpcr-target-biology.
Catalog No. 50-200-5957 Supplier Eurofins Discoverx Supplier No. HTS137C
Add to Cart
Add to Cart
Request a Quote
The ChemiScreen CaS Calcium Sensor Receptor Stable Cell Line is an engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. extracellular calcium) based activation of CASR GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses CASR alongside endogenous high levels of the promiscuous G protein, Gα15, coupling the receptor to the calcium signaling pathway. Upon activation of CASR, the resulting calcium activity can be fluorescently assayed. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, visit www.discoverx.com/targets/gpcr-target-biology.