Learn More
TARGETMOL CHEMICALS INC CASIN 10MG
SDP
Catalog No. 502576851
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CASIN (Pirl1-related Compound 2) is a specific inhibitor of GTPase Cdc42 (IC50 2 uM). purity: 99%
Catalog No. 50-257-6851 Supplier TARGETMOL CHEMICALS INC Supplier No. T397110MG
Add to Cart
Add to Cart
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CASIN (Pirl1-related Compound 2) is a specific inhibitor of GTPase Cdc42 (IC50 2 uM). purity: 99%