Learn More
NEOGEN Casman Medium Base, 500 g
SDP
Catalog No. 502015558
Encompass_Preferred
Casman Medium Base is used with blood for the isolation of Haemophilus influenzae, Neisseria gonorrhoeae and other fastidious microorganisms in a laboratory setting. Casman Medium Base is not intended for use in the diagnosis of disease or other conditions in humans.In 1947, Casman described an infusion-free medium enriched with 5% blood for fastidious microorganisms incubated anaerobically. This medium replaced formulas containing fresh meat infusion and heated blood. Casman adjusted the formula after experiments revealed nicotinamide disrupted the action of a blood enzyme that inactivates V factor (NAD). The concentration of nicotinamide was lowered to support growth of Neisseria spp.For more information visit https://www.neogen.com/solutions/Microbiology/casman-medium-base
Catalog No. 50-201-5558 Supplier NEOGEN Supplier No. 700004531
Casman Medium Base is used with blood for the isolation of Haemophilus influenzae, Neisseria gonorrhoeae and other fastidious microorganisms in a laboratory setting. Casman Medium Base is not intended for use in the diagnosis of disease or other conditions in humans.In 1947, Casman described an infusion-free medium enriched with 5% blood for fastidious microorganisms incubated anaerobically. This medium replaced formulas containing fresh meat infusion and heated blood. Casman adjusted the formula after experiments revealed nicotinamide disrupted the action of a blood enzyme that inactivates V factor (NAD). The concentration of nicotinamide was lowered to support growth of Neisseria spp.For more information visit https://www.neogen.com/solutions/Microbiology/casman-medium-base