Learn More
Medchemexpress LLC Casopitant mesylate | 414910-30-8 | 99.1% | C31H39F7N4O5S | 50 MG
SDP
Catalog No. 5000355784
Encompass_Preferred
Casopitant mesylate (GW679769B) is a potent, selective, brain-permeable, and orally active neurokinin 1 (NK1) receptor antagonist. It is classified as a second-in-class antiemetic that counteracts the emetogenic effect of Substance P. It is utilized for the treatment of chemotherapy-induced nausea and vomiting (CINV) and postoperative nausea and vomiting (PONV).
  • Potent, selective, brain-permeable, and orally active neurokinin 1 (NK1) receptor antagonist.
  • Antagonizes the emetogenic effect of Substance P.
  • Substrate and a weak-to-moderate inhibitor of CYP3A4.
  • Used for the treatment of chemotherapy-induced nausea and vomiting (CINV).
  • Used for the treatment of postoperative nausea and vomiting (PONV).
Catalog No. 50-003-55784 Supplier Medchemexpress LLC Supplier No. HY14405A50MG
Casopitant mesylate (GW679769B) is a potent, selective, brain-permeable, and orally active neurokinin 1 (NK1) receptor antagonist. It is classified as a second-in-class antiemetic that counteracts the emetogenic effect of Substance P. It is utilized for the treatment of chemotherapy-induced nausea and vomiting (CINV) and postoperative nausea and vomiting (PONV).
  • Potent, selective, brain-permeable, and orally active neurokinin 1 (NK1) receptor antagonist.
  • Antagonizes the emetogenic effect of Substance P.
  • Substrate and a weak-to-moderate inhibitor of CYP3A4.
  • Used for the treatment of chemotherapy-induced nausea and vomiting (CINV).
  • Used for the treatment of postoperative nausea and vomiting (PONV).