Learn More
Medchemexpress LLC Casopitant mesylate | 414910-30-8 | 99.8% | 712.72 g/mol | C31H39F7N4O5S | 5 MG
SDP
Catalog No. 5000221401
Encompass_Preferred
Casopitant mesylate is a research-grade neurokinin-1 (NK1) receptor antagonist supplied as the mesylate salt for laboratory studies. It is used in pharmacological and biochemical research on antiemetic activity and CNS NK1 receptor signaling, available in solid and solution formats and characterized by standard analytical methods.
  • Potent NK1 receptor antagonist for receptor and pharmacology studies.
  • High purity suitable for analytical and in vitro applications.
  • Available in multiple solid pack sizes and ready-to-use solution formats.
  • Characterized by RP-HPLC, NP-HPLC, and LCMS for quality verification.
  • Intended for research use only; not for human or clinical use.
Catalog No. 50-002-21401 Supplier Medchemexpress LLC Supplier No. HY14405A5MG
Casopitant mesylate is a research-grade neurokinin-1 (NK1) receptor antagonist supplied as the mesylate salt for laboratory studies. It is used in pharmacological and biochemical research on antiemetic activity and CNS NK1 receptor signaling, available in solid and solution formats and characterized by standard analytical methods.
  • Potent NK1 receptor antagonist for receptor and pharmacology studies.
  • High purity suitable for analytical and in vitro applications.
  • Available in multiple solid pack sizes and ready-to-use solution formats.
  • Characterized by RP-HPLC, NP-HPLC, and LCMS for quality verification.
  • Intended for research use only; not for human or clinical use.