Learn More
Medchemexpress LLC Casopitant mesylate | 414910-30-8 | 99.8% | C31H39F7N4O5S | 10MG
SDP
Catalog No. 5000259138
Encompass_Preferred
Casopitant mesylate is a potent, selective, brain-permeable, and orally active neurokinin-1 (NK1) receptor antagonist supplied as the mesylate salt for preclinical research into antiemetic mechanisms, including chemotherapy-induced and postoperative nausea and vomiting. The compound is offered with high reported purity and in multiple formats for laboratory use.
  • Potent, selective NK1 receptor antagonist
  • Brain permeable and orally active
  • Supplied as mesylate salt for improved handling
  • High reported purity suitable for research
  • Available in multiple sizes and solution formats
  • Intended for preclinical antiemetic and pharmacology studies
Catalog No. 50-002-59138 Supplier Medchemexpress LLC Supplier No. HY14405A10MG
Casopitant mesylate is a potent, selective, brain-permeable, and orally active neurokinin-1 (NK1) receptor antagonist supplied as the mesylate salt for preclinical research into antiemetic mechanisms, including chemotherapy-induced and postoperative nausea and vomiting. The compound is offered with high reported purity and in multiple formats for laboratory use.
  • Potent, selective NK1 receptor antagonist
  • Brain permeable and orally active
  • Supplied as mesylate salt for improved handling
  • High reported purity suitable for research
  • Available in multiple sizes and solution formats
  • Intended for preclinical antiemetic and pharmacology studies