Learn More
CPC Scientific Ac-Leu-Glu-His-Asp-AFC 1MG
SDP
Catalog No. 502109084
Encompass
SEQUENCE: Ac-Leu-Glu-His-Asp-AFCONE-LETTER SEQUENCE: Ac-LEHD-AFCMOLECULAR FORMULA: C33H38F3N7O11MOLECULAR WEIGHT: 765.7STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [210345-03-2]SYNONYMS: N-Acetyl-Leu-Glu-His-Asp-7-amino-4-Trifluoromethylcoumarin, Caspase-9 substrate (Fluorogenic)RESEARCH AREA: ApoptosisCITATIONS:Ribeiro, V.; Ferreres, F., et al. Food Research International 2021, 141, 110121.?REFERENCES:R.Moriya et al., FEBS Lett. , 484, 253 (2000)SKU(s): CASP-088A, CASP-088B
Catalog No. 50-210-9084 Supplier CPC Scientific Supplier No. CASP088A
SEQUENCE: Ac-Leu-Glu-His-Asp-AFCONE-LETTER SEQUENCE: Ac-LEHD-AFCMOLECULAR FORMULA: C33H38F3N7O11MOLECULAR WEIGHT: 765.7STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [210345-03-2]SYNONYMS: N-Acetyl-Leu-Glu-His-Asp-7-amino-4-Trifluoromethylcoumarin, Caspase-9 substrate (Fluorogenic)RESEARCH AREA: ApoptosisCITATIONS:Ribeiro, V.; Ferreres, F., et al. Food Research International 2021, 141, 110121.?REFERENCES:R.Moriya et al., FEBS Lett. , 484, 253 (2000)SKU(s): CASP-088A, CASP-088B