Learn More
Apexbio Technology LLC Caspase-3-Fluoro-Assay-Kit
SDP
Catalog No. 502240287
Encompass_Preferred
Caspase-3 Fluorometric Assay Kit is widely used in biomedical research to study cell apoptosis Originating from studies on apoptosis pathways this kit detects the activity of caspase-3 an enzyme playing a critical role in the execution phase of cell apoptosis It targets the cleavage of the specific substrate releasing a fluorescent reporter that allows quantification of enzyme activity This detection method is highly sensitive enabling the measurement of caspase activity in the nanomolar to low micromolar range Commonly applied in vitro it is essential for evaluating programmed cell death in various cell models including cancer and stem cells with applications extending to animal studies and drug screening Experimental concentrations typically depend on the specific research design providing flexibility in optimizing conditions for diverse experimental setups and requirements
This item is not returnable. View return policy
Catalog No. 50-224-0287 Supplier Apexbio Technology LLC Supplier No. K200725
Caspase-3 Fluorometric Assay Kit is widely used in biomedical research to study cell apoptosis Originating from studies on apoptosis pathways this kit detects the activity of caspase-3 an enzyme playing a critical role in the execution phase of cell apoptosis It targets the cleavage of the specific substrate releasing a fluorescent reporter that allows quantification of enzyme activity This detection method is highly sensitive enabling the measurement of caspase activity in the nanomolar to low micromolar range Commonly applied in vitro it is essential for evaluating programmed cell death in various cell models including cancer and stem cells with applications extending to animal studies and drug screening Experimental concentrations typically depend on the specific research design providing flexibility in optimizing conditions for diverse experimental setups and requirements