Learn More
Enzo Life Sciences Caspase-9 (full-length) (human), (recombinant) (50 kU)
SDP
Catalog No. 502011958
Encompass_Preferred
Alternative name: ICE-LAP-6, Mch6. Concentration: 500 U/µl. Purity: ≥85% (SDS-PAGE). Formulation: Liquid. In 50 mM HEPES, pH 7.4, 100mM NaCl, 0.5% CHAPS, 1mM EDTA, 10% glycerol, 10mM DTT. Source: Produced in E. coli. Full length human caspase-9 (aa 1-416). Long Term Storage: -80°C.
Catalog No. 50-201-1958 Supplier Enzo Life Sciences Supplier No. BMLSE2400050
Alternative name: ICE-LAP-6, Mch6. Concentration: 500 U/µl. Purity: ≥85% (SDS-PAGE). Formulation: Liquid. In 50 mM HEPES, pH 7.4, 100mM NaCl, 0.5% CHAPS, 1mM EDTA, 10% glycerol, 10mM DTT. Source: Produced in E. coli. Full length human caspase-9 (aa 1-416). Long Term Storage: -80°C.