Learn More
Enzo Life Sciences Caspase-fluorometric substrate set (1 Set)
SDP
Catalog No. 502010121
Encompass_Preferred
Fluorometric substrates for assaying activities of members of caspase family proteases. All substrates are provided in liquid ready-to-use form. Long Term Storage: -20°C. Applications: Fluorescence microscopy .
Catalog No. 50-201-0121 Supplier Enzo Life Sciences Supplier No. ALX850229KI01
Fluorometric substrates for assaying activities of members of caspase family proteases. All substrates are provided in liquid ready-to-use form. Long Term Storage: -20°C. Applications: Fluorescence microscopy .