Learn More
Aviva Systems Biology Corp CASP1 Antibody - middle region (ARP58983_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58983_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CASP1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human CASP1. Alternate names: ICE, IL1BC, P45ApplicationWB. Species reactivity: Horse, Human, Rabbit, Yeast
Catalog No. 50-286-549