Learn More
Aviva Systems Biology Corp CASP5 Antibody - middle region (ARP58992_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58992_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CASP5. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human CASP5. Alternate names: ICE(rel)III, ICEREL-III, ICH-3, MGC141966ApplicationWB. Species reactivity: Cow, Guinea Pig, Human
Catalog No. 50-286-552