Learn More
Aviva Systems Biology Corp CASP6 Antibody - middle region (ARP58994_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58994_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CASP6. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human CASP6. Alternate names: MCH2ApplicationWB. Species reactivity: Human, Mouse, Pig
Catalog No. 50-286-553