Learn More
Aviva Systems Biology Corp CASP7 Antibody - N-terminal region (ARP58995_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58995_P050
Encompass_Preferred
To order by phone call us at (888) 880-0001, fax us at (858) 552-6975 or send an email to info@avivasysbio.com. Aviva manufactures this antibody so we can offer the best price. Please contact us to request pricing information.
All of Aviva’s products are guaranteed.r the applications and experimental sample types mentioned in the datasheet below. Are you curious if this product will work.r you? Please contact us at techsupport@avivasysbio.com
The immunogen is a synthetic peptide directed towards the N terminal region of human CASP7. Alternate names: CMH-1, ICE-LAP3, MCH3ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-286-554