Learn More
Diagnostic Biosystems CASPASE 1 Caspase 1/Interleukin-1-β Converting Enzyme
Catalog No. 502407401
Encompass_Preferred
Caspase 1 and is expressed as a proenzyme in many tissues. Active Caspase 1 is a tetramer of two subunits p10 and p20 in 2:2 ratio. Overexpression of caspase 1 can induce apoptosis in fibroblast, which can be inhibited by overexpression of Crm A, a protein from pox virus.
Catalog No. 50-240-7401 Supplier Diagnostic Biosystems Supplier No. PDR175
Caspase 1 and is expressed as a proenzyme in many tissues. Active Caspase 1 is a tetramer of two subunits p10 and p20 in 2:2 ratio. Overexpression of caspase 1 can induce apoptosis in fibroblast, which can be inhibited by overexpression of Crm A, a protein from pox virus.