Learn More
Santa Cruz Biotechnology Caspase-1 Antibody (14F468), 100 ug/mL
Supplier:  Santa Cruz Biotechnology SC56036
Encompass
Recommended for detection of full-length caspase-1 and cleaved caspase-1 forms that retain amino acids 371-390 of the caspase-1 protein of mouse, rat, human and porcine origin by WB, IP, IF and IHC(P)
Catalog No. NC1830105