Learn More
Enzo Life Sciences Caspase-1 (human), (recombinant) (active) (25U)
SDP
Supplier:  Enzo Life Sciences ALX201056U025
Encompass_Preferred
Alternative name: Interleukin 1 beta converting enzyme, IL-1 beta converting enzyme, ICE. Formulation: Lyophilized. Source: Produced in E. coli. Contains an N-terminal His-tag. Long Term Storage: -80°C. Handling: Avoid freeze/thaw cycles. After reconstitution, prepare aliquots and store at -80°C.
Catalog No. 50-200-7898