Learn More
CPC Scientific Ac-Ala-Ala-Val-Ala-Leu-Leu-Pro-Ala-Val-Leu-Leu-Ala-Leu-Leu-Ala-Pro-Tyr-Val-Ala-Asp-CHO 1MG
SDP
Catalog No. 502108930
Encompass
Cell permeable inhibitor of caspase-1.ONE-LETTER SEQUENCE: Ac-AAVALLPAVLLALLAPYVAD-CHOMOLECULAR FORMULA: C97H160N20O24MOLECULAR WEIGHT:1990.46STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [201608-12-0], SYNONYMS: Cell-permeable Caspase-1 Inhibitor IRESEARCH AREA: ApoptosisREFERENCES: Y.-Z. Lin et al., J. Biol. Chem., 271, 5305 (1996); L.A. Reiter, Int. J. Pept. Protein Res., 43, 87 (1994); N.A. Thornberry et al., Biochemistry, 33, 3934 (1994)
Catalog No. 50-210-8930 Supplier CPC Scientific Supplier No. CASP001B
Cell permeable inhibitor of caspase-1.ONE-LETTER SEQUENCE: Ac-AAVALLPAVLLALLAPYVAD-CHOMOLECULAR FORMULA: C97H160N20O24MOLECULAR WEIGHT:1990.46STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [201608-12-0], SYNONYMS: Cell-permeable Caspase-1 Inhibitor IRESEARCH AREA: ApoptosisREFERENCES: Y.-Z. Lin et al., J. Biol. Chem., 271, 5305 (1996); L.A. Reiter, Int. J. Pept. Protein Res., 43, 87 (1994); N.A. Thornberry et al., Biochemistry, 33, 3934 (1994)