Learn More
Enzo Life Sciences Caspase-1 (mouse) monoclonal antibody (1H11) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX804530C100
Encompass_Preferred
Clone: 1H11. Isotype: IgG1. Immunogen: p20 fragment of mouse caspase-1 and synthetic peptide corresponding to aa 206-220 (T206ALEMVKEVKEFAAC220) of mouse caspase-1. Species reactivity: Mouse. Specificity: Recognizes an epitope within the p20 fragment of caspase-1. Applications: ELISA, Flow Cytometry, WB. Formulation: Liquid. Protein G Sepharose™ purified antibody in PBS containing 0.02% sodium azide.
Catalog No. 50-200-9937