Learn More
Enzo Life Sciences Caspase-1 polyclonal antibody (100µg)
SDP
Supplier:  Enzo Life Sciences ALX210804C100
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to aa 224-242 (CEPH224KTSDSTFLVFMSHGIREG242) of human caspase-1. Species reactivity: Human, Mouse, Rat. Applications: ICC, IHC, IP, WB. Purity: Epitope-affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 30% glycerol, 0.5% BSA and 0.01% thimerosal.
Catalog No. 50-200-8161