Learn More
Echelon Biosciences Research Labs Caspase-1 Substrate - 1 mg, CAS#: 189696-01-3
SDP
Catalog No. 501908937
Encompass_Preferred
Caspase-1 Substrate - 1mg , Mca-Tyr-Val-Ala-Asp-Ala-Pro-Lys(DNP)-OH Category/Application: Enzyme Substrates This item is non returnable
This item is not returnable. View return policy
Catalog No. 50-190-8937 Supplier Echelon Biosciences Research Labs Supplier No. 865211MG
Caspase-1 Substrate - 1mg , Mca-Tyr-Val-Ala-Asp-Ala-Pro-Lys(DNP)-OH Category/Application: Enzyme Substrates This item is non returnable