Learn More
Enzo Life Sciences Caspase 10 polyclonal antibody (100 µg)
SDP
Supplier:  Enzo Life Sciences ADIAAP110E
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to aa 505-521 of human Caspase-10. UniProt: Q92851. Species Reactivity: Human. Applications: WB. Recommended dilutions: Western Blot (2µg/ml, ECL). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: DEAE purified. Formulation: Liquid. In PBS containing 0.02% sodium azide.
Catalog No. 50-658-749