Learn More
AdipoGen Caspase-11 ms mAb 8A5
SDP
Supplier:  AdipoGen AG20T0139C100
Encompass_Preferred
Monoclonal Antibody. Recognizes mouse and rat full-length caspase-11 protein. Detects two bands of ~38kDa and ~43kDa by Western blot. Isotype Rat IgG1kappa. Clone 8A5. Applications ELISA, FACS, IP, WB. Liquid. In PBS containing 0.02% sodium azide. Caspase-11 not only activates caspase-1, that is required for the maturation of proinflammatory cytokines such as interleukin-1 IL-1 and interleukin-18 IL-18, but also activates caspase-3, leading to cellular apoptosis under pathological conditions. In most cells, caspase-11 is only expressed upon induction with pro-inflammatory stimuli. Caspase-11 is an essential mediator of endotoxic shock and caspase-11-deficient mice are resistant to endotoxic shock. LPS-induced caspase-11 regulates lymphocyte apoptosis by activating both caspase-3 and caspase-7.
Catalog No. 50-256-3341