Learn More
Enzo Life Sciences Caspase-11 (mouse) monoclonal antibody (8A5) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX804494C100
Encompass_Preferred
Clone: 8A5. Isotype: IgG1. Immunogen: p20 fragment of mouse caspase-11. Species reactivity: Mouse, Rat. Specificity: The epitope is in the p20 fragment. Applications: Flow Cytometry, WB. Purity: Protein G-affinity purified. Formulation: Liquid. In PBS containing 0.02% sodium azide.
Catalog No. 50-200-9923