Learn More
Apexbio Technology LLC Caspase-2-Fluoro-Assay-Kit
SDP
Catalog No. 5000567754
Encompass_Preferred
Caspase-2 Fluorometric Assay Kit is used in biomedical research for detecting caspase-2 activity an important target in apoptosis pathways The assay originates from studies focusing on programmed cell death helping to understand its role in both normal cellular processes and diseases Caspase-2 is an initiator caspase that responds to apoptotic stimuli and plays a critical role in activating downstream effectors The kit measures in vitro activity typically ranging from nanomolar to low micromolar providing important insights into apoptotic pathways It is widely applied in cell models animal studies and drug screening to evaluate apoptosis-related effects Experimental concentrations typically depend on the specific research design This kit aids researchers in studying potential therapeutic agents targeting apoptotic pathways contributing to the development of treatments for diseases such as cancer and neurodegenerative disorders
Catalog No. 50-005-67754 Supplier Apexbio Technology LLC Supplier No. K2016400
Add to Cart
Add to Cart
Caspase-2 Fluorometric Assay Kit is used in biomedical research for detecting caspase-2 activity an important target in apoptosis pathways The assay originates from studies focusing on programmed cell death helping to understand its role in both normal cellular processes and diseases Caspase-2 is an initiator caspase that responds to apoptotic stimuli and plays a critical role in activating downstream effectors The kit measures in vitro activity typically ranging from nanomolar to low micromolar providing important insights into apoptotic pathways It is widely applied in cell models animal studies and drug screening to evaluate apoptosis-related effects Experimental concentrations typically depend on the specific research design This kit aids researchers in studying potential therapeutic agents targeting apoptotic pathways contributing to the development of treatments for diseases such as cancer and neurodegenerative disorders