Learn More
Enzo Life Sciences Caspase-2 fluorometric assay kit (25 tests)
SDP
Catalog No. 502010111
Encompass_Preferred
Based on the detection of AFC (7-amino-4-trifluoromethyl coumarin) after cleavage from the labelled substrate VDVAD-AFC. Applications: Fluorescent detection Activity assay.
Catalog No. 50-201-0111 Supplier Enzo Life Sciences Supplier No. ALX850214KI01
Based on the detection of AFC (7-amino-4-trifluoromethyl coumarin) after cleavage from the labelled substrate VDVAD-AFC. Applications: Fluorescent detection Activity assay.