Learn More
AdipoGen Caspase-2 mAb 11B4
SDP
Supplier:  AdipoGen AG20T0136C100
Encompass_Preferred
Monoclonal Antibody. Recognizes an epitope in the p19 subunit of human, mouse, rat, monkey and dog caspase-2. Detects bands of ~51kDa procaspase-2, ~32kDa and ~18kDa cleavage products by Western blot. Isotype Rat IgG2akappa. Clone 11B4. Applications ELISA, FACS, ICC, IHC, IP, WB. Liquid. In PBS containing 0.02% sodium azide. Caspase-2 is a Class I caspase with a long prodomain necessary for nuclear localization. Upon activation of the apoptotic pathway, the procaspase is cleaved into smaller fragments. Caspase-2 is the nuclear apoptotic respondent to cellular genotoxic stress or mitotic catastrophe and is involved in the activation cascade of caspases responsible for apoptosis execution. Activation occurs upon recruitment to a complex containing a p53-induced death domain protein, PIDD. This suggests caspase-2 can be a nuclear initiator caspase with a requirement for caspase-9 and caspase-3 activation in downstream apoptotic events.
Catalog No. 50-256-3343