Learn More
Enzo Life Sciences Caspase-2 monoclonal antibody (11B4) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX804356C100
Encompass_Preferred
Clone: 11B4. Isotype: IgG2a. Immunogen: Recombinant human caspase-2 p19 fragment. Species reactivity: Human, Mouse, Rat, Monkey, Dog. Specificity: Recognizes an epitope in the p19 subunit of human, mouse, rat, monkey and dog caspase-2. Detects bands of ~51kDa (procaspase-2), ~32kDa and ~18kDa (cleavage products) by Western blot. Applications: IP, WB. Purity: Protein G-affinity purified. Formulation: Liquid. In PBS containing 0.02% sodium azide.
Catalog No. 50-200-9881