Learn More
Enzo Life Sciences Caspase-2 monoclonal antibody (4-1-5) (100µg)
SDP
Supplier:  Enzo Life Sciences ADI905685100
Encompass_Preferred
Clone: 4-1-5. Host: Mouse. Isotype: IgG1. Immunogen: Recombinant full length human Caspase-2. Source: Purified from mouse ascites. Species reactivity: Human. Applications: WB. Purity: Protein G affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 0.09% sodium azide and 50% glycerol.
Catalog No. 50-200-6993