Learn More
Enzo Life Sciences Caspase-3 (human), (recombinant) (active) (100 U)
SDP
Supplier:  Enzo Life Sciences ALX201059U100
Encompass_Preferred
Alternative name: CPP32, Yama, Apopain. Formulation: Lyophilized. Source: Produced in E. coli. Contains an N-terminal His-tag. UniProt: P42574. Handling: Avoid freeze/thaw cycles. After reconstitution, prepare aliquots and store at -80°C. Long Term Storage: -80°C
Catalog No. NC1332378