Learn More
STEMCELL Technologies Caspase-3/7 Activity Flow Cytometer Kit, Green, 100 Tests
SDP
Catalog No. NC3823368
Encompass
Efficiently detect active caspase-3 in mammalian cells undergoing apoptosis by using the Caspase-3/7 Activity Flow Cytometry Kit, Green. This reagent kit uses a quick and simple staining protocol that is highly sensitive and compatible with standard flow cytometers.Caspase-3 and caspase-7 have a proteolytic substrate selectivity for the peptide sequence Asp-Glu-Val-Asp (DEVD). This kit includes TF2-DEVD-FMK—a cell-permeable and non-toxic fluorescent substrate—as well as an assay buffer and propidium iodide. When added to apoptotic cells, TF2-DEVD-FMK irreversibly binds to activated caspase-3, resulting in the retention of the green fluorescent reagent within cells that can be detected by flow cytometry.
Catalog No. NC3823368 Supplier STEMCELL Technologies Supplier No. 1000923
Efficiently detect active caspase-3 in mammalian cells undergoing apoptosis by using the Caspase-3/7 Activity Flow Cytometry Kit, Green. This reagent kit uses a quick and simple staining protocol that is highly sensitive and compatible with standard flow cytometers.Caspase-3 and caspase-7 have a proteolytic substrate selectivity for the peptide sequence Asp-Glu-Val-Asp (DEVD). This kit includes TF2-DEVD-FMK—a cell-permeable and non-toxic fluorescent substrate—as well as an assay buffer and propidium iodide. When added to apoptotic cells, TF2-DEVD-FMK irreversibly binds to activated caspase-3, resulting in the retention of the green fluorescent reagent within cells that can be detected by flow cytometry.