Learn More
Apexbio Technology LLC Caspase-3/7 Inhibitor I,1mg CAS# 220509-74-0
SDP
Catalog No. 502240000
Encompass_Preferred
Caspase-3/7 inbibitor I is a potent, reversible, isatin sulfonamide-based inhibitor of caspase-3 (KI(app) = 60 nM) and caspase-7 (KI(app) = 170 nM). Is a weaker inhibitor of caspase-9 (Ki(app) = 3.1 mM). It Has only a trivial effect (Ki(app) >25 mM) on the activities of caspase-1, caspase-2, caspase-4, caspase-6, and caspase-8. It has been shown to inhibit apoptosis in camptothecin treated Jurkat cells (IC50 ~50 µM). Also it has been reported to inhibit apoptosis in chondrocytes (44% inhibition at 10 µM and 98% inhibition at 50 µM). Selectivity for caspases-3 and 7 involves unique hydrophobic residues in the S2 pocket surrounding the catalytic cysteine residue. [1] [2] In some systems inhibition of caspases-3 and -7 can prevent apoptosis and may therefore have important therapeutic implications. Other sizes are also available. Please inqury us for quote.
This item is not returnable. View return policy
Catalog No. 50-224-0000 Supplier Apexbio Technology LLC Supplier No. A19251
Caspase-3/7 inbibitor I is a potent, reversible, isatin sulfonamide-based inhibitor of caspase-3 (KI(app) = 60 nM) and caspase-7 (KI(app) = 170 nM). Is a weaker inhibitor of caspase-9 (Ki(app) = 3.1 mM). It Has only a trivial effect (Ki(app) >25 mM) on the activities of caspase-1, caspase-2, caspase-4, caspase-6, and caspase-8. It has been shown to inhibit apoptosis in camptothecin treated Jurkat cells (IC50 ~50 µM). Also it has been reported to inhibit apoptosis in chondrocytes (44% inhibition at 10 µM and 98% inhibition at 50 µM). Selectivity for caspases-3 and 7 involves unique hydrophobic residues in the S2 pocket surrounding the catalytic cysteine residue. [1] [2] In some systems inhibition of caspases-3 and -7 can prevent apoptosis and may therefore have important therapeutic implications. Other sizes are also available. Please inqury us for quote.