Learn More
Echelon Biosciences Research Labs Caspase 3 Apopain Substrate - 5 mg, CAS#: 201608-14-2
SDP
Supplier:  Echelon Biosciences Research Labs 863175MG
Encompass_Preferred
Caspase 3 Apopain Substrate - 5mg , Ac-Asp-Glu-Val-Asp-AFC Category/Application: Enzyme Substrates This item is non returnable
Catalog No. 50-190-8902