Learn More
Sigma Aldrich Fine Chemicals Biosciences Caspase 3 Assay Kit Fluori
Catalog No. 502907814
Encompass
The Caspase 3 Fluorometric Assay Kit is based on the hydrolysis of acetyl Asp-Glu-Val-Asp 7-amido-4-methylcoumarin (Ac-DEVD-AMC) by caspase 3 resulting in the release of the fluorescent 7-amino-4-methylcoumarin (AMC). The excitation and emission wavelengths of AMC are 360 nm and 460 nm respectively. The concentration of the AMC released can be calculated from a calibration curve prepared with AMC standards (AMC standard included with the kit).
Catalog No. 50-290-7814 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. CASP3F1KT
The Caspase 3 Fluorometric Assay Kit is based on the hydrolysis of acetyl Asp-Glu-Val-Asp 7-amido-4-methylcoumarin (Ac-DEVD-AMC) by caspase 3 resulting in the release of the fluorescent 7-amino-4-methylcoumarin (AMC). The excitation and emission wavelengths of AMC are 360 nm and 460 nm respectively. The concentration of the AMC released can be calculated from a calibration curve prepared with AMC standards (AMC standard included with the kit).