Learn More
Cell Signaling Technology Caspase-3 (D3R6Y) Rabbit Monoclonal Antibody, Recombinant, Unconjugated, 100 μL
SDP
Catalog No. NC1573999
Encompass
This rabbit monoclonal antibody is designed to detect endogenous levels of total caspase-3 protein, specifically recognizing both the full-length protein and the large subunit (p20) generated during proteolytic cleavage in apoptosis. The antibody is produced by immunizing animals with recombinant protein specific to the human caspase-3 p20 subunit.
  • Validated for western blotting, simple western, and immunoprecipitation applications.
  • Reacts with human, mouse, rat, and monkey species, with predicted reactivity in pig.
  • Supplied in a stable formulation containing 10 mM sodium HEPES (pH 7.5), 150 mM NaCl, 100 μg/mL BSA, 50% glycerol, and <0.02% sodium azide.
  • Recommended storage at -20°C without aliquoting.
Catalog No. NC1573999 Supplier Cell Signaling Technology Supplier No. 14220S
This rabbit monoclonal antibody is designed to detect endogenous levels of total caspase-3 protein, specifically recognizing both the full-length protein and the large subunit (p20) generated during proteolytic cleavage in apoptosis. The antibody is produced by immunizing animals with recombinant protein specific to the human caspase-3 p20 subunit.
  • Validated for western blotting, simple western, and immunoprecipitation applications.
  • Reacts with human, mouse, rat, and monkey species, with predicted reactivity in pig.
  • Supplied in a stable formulation containing 10 mM sodium HEPES (pH 7.5), 150 mM NaCl, 100 μg/mL BSA, 50% glycerol, and <0.02% sodium azide.
  • Recommended storage at -20°C without aliquoting.