Learn More
Enzo Life Sciences Caspase-3 fluorometric assay kit (25 tests)
SDP
Catalog No. 502010113
Encompass_Preferred
Based on the detection of AFC (7-amino-4-trifluoromethyl coumarin) after cleavage from the labelled substrate DEVD-AFC. Alternative name: CPP32, Yama, Apopain. Applications: Fluorescent detection Activity assay.
Catalog No. 50-201-0113 Supplier Enzo Life Sciences Supplier No. ALX850216KI01
Based on the detection of AFC (7-amino-4-trifluoromethyl coumarin) after cleavage from the labelled substrate DEVD-AFC. Alternative name: CPP32, Yama, Apopain. Applications: Fluorescent detection Activity assay.