Learn More
Enzo Life Sciences Caspase-3 (human), (recombinant) (active) (25U)
SDP
Catalog No. 502007901
Encompass_Preferred
Alternative name: CPP32, Yama, Apopain. Formulation: Lyophilized. Source: Produced in E. coli. Contains an N-terminal His-tag. Long Term Storage: -80°C.
Catalog No. 50-200-7901 Supplier Enzo Life Sciences Supplier No. ALX201059U025
Alternative name: CPP32, Yama, Apopain. Formulation: Lyophilized. Source: Produced in E. coli. Contains an N-terminal His-tag. Long Term Storage: -80°C.