Learn More
Sigma Aldrich Fine Chemicals Biosciences Caspase 3 human 90 SDS PAG
Catalog No. 502905473
Encompass
Human recombinant C-terminal histidine tagged caspase 3 is a fully active protein consisting of 17 kDa and 13.5 kDa subunits the 13.5 kDa subunit contains the histidine tag.
Catalog No. 50-290-5473 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. C122410UG
Human recombinant C-terminal histidine tagged caspase 3 is a fully active protein consisting of 17 kDa and 13.5 kDa subunits the 13.5 kDa subunit contains the histidine tag.