Learn More
Enzo Life Sciences Caspase-3 (human), (recombinant) (5000 U)
SDP
Catalog No. NC9989464
Encompass_Preferred
Alternative name: CPP32. Yama. Apopain. MW: 17 + 12 kDa. Concentration: 100 U/µl. Purity: ≥95% (SDS-PAGE). Formulation: Liquid. In 50mM HEPES, pH 7.4, 100mM sodium chloride, 0.5% CHAPS, 1mM EDTA, 10% glycerol and 10mM dithiothreitol. Source: Produced in E. coli. Caspase-3 (CPP32; Yama; apopain) from human cDNA. The enzyme was cleaved and activated from the proenzyme UniProt: P42574. Handling: Avoid freeze/thaw cycles. Long Term Storage: -80°C
Catalog No. NC9989464 Supplier Enzo Life Sciences Supplier No. BMLSE1695000
Alternative name: CPP32. Yama. Apopain. MW: 17 + 12 kDa. Concentration: 100 U/µl. Purity: ≥95% (SDS-PAGE). Formulation: Liquid. In 50mM HEPES, pH 7.4, 100mM sodium chloride, 0.5% CHAPS, 1mM EDTA, 10% glycerol and 10mM dithiothreitol. Source: Produced in E. coli. Caspase-3 (CPP32; Yama; apopain) from human cDNA. The enzyme was cleaved and activated from the proenzyme UniProt: P42574. Handling: Avoid freeze/thaw cycles. Long Term Storage: -80°C