Learn More
CPC Scientific Ac-Ala-Ala-Val-Ala-Leu-Leu-Pro-Ala-Val-Leu-Leu-Ala-Leu-Leu-Ala-Pro-Asp-Glu-Val-Asp-CHO 1MG
SDP
Catalog No. 502109003
Encompass
ONE-LETTER SEQUENCE: Ac-AAVALLPAVLLALLAPDEVD-CHOMOLECULAR FORMULA: C94H158N20O27MOLECULAR WEIGHT:2000.4STORAGE CONDITIONS: -20 5CRESEARCH AREA: ApoptosisREFERENCES: Y.Z. Lin et al., J. Biol. Chem., 271, 5305 (1996); D.W. Nicholson, Nature Biotechnol., 14, 297 (1996); M. Tewari et al., Cell, 81, 801 (1995)
Catalog No. 50-210-9003 Supplier CPC Scientific Supplier No. CASP047B
ONE-LETTER SEQUENCE: Ac-AAVALLPAVLLALLAPDEVD-CHOMOLECULAR FORMULA: C94H158N20O27MOLECULAR WEIGHT:2000.4STORAGE CONDITIONS: -20 5CRESEARCH AREA: ApoptosisREFERENCES: Y.Z. Lin et al., J. Biol. Chem., 271, 5305 (1996); D.W. Nicholson, Nature Biotechnol., 14, 297 (1996); M. Tewari et al., Cell, 81, 801 (1995)