Learn More
Cayman Chemical Caspase-3 Inhibitor VII, Purity: ≥98%, CAS: 745046-84-8, 1 mg
SDP
Catalog No. 502952005
Encompass_Preferred
A reversible caspase-3 inhibitor (IC50 = 23 nM); displays anti-apoptotic activity in Jurkat T cells treated with staurosporine
Catalog No. 50-295-2005 Supplier Cayman Chemical Supplier No. 133061 MG
A reversible caspase-3 inhibitor (IC50 = 23 nM); displays anti-apoptotic activity in Jurkat T cells treated with staurosporine