Learn More
Cayman Chemical Caspase-3 InhIbItr VII 5mg
SDP
Catalog No. 502952543
Encompass_Preferred
A reversible caspase-3 inhibitor (IC50 = 23 nM); displays anti-apoptotic activity in Jurkat T cells treated with staurosporine
Catalog No. 50-295-2543 Supplier Cayman Chemical Supplier No. 133065 MG
A reversible caspase-3 inhibitor (IC50 = 23 nM); displays anti-apoptotic activity in Jurkat T cells treated with staurosporine