Learn More
Bioss Caspase-3 - N-terminal region
SDP
Catalog No. 5000065153
Encompass_Preferred
Caspase-3 (N-terminal region) Primary Antibody Unconjugated Host Species Mouse Target Species Human Clonality Monoclonal Applications WB M377Concentration Size 100 ul Storage Buffer PBS 1 mg/ml BSA 0 05% NaN3 and 50% glycerol
Catalog No. 50-000-65153 Supplier Bioss Supplier No. BSM70373M
Caspase-3 (N-terminal region) Primary Antibody Unconjugated Host Species Mouse Target Species Human Clonality Monoclonal Applications WB M377Concentration Size 100 ul Storage Buffer PBS 1 mg/ml BSA 0 05% NaN3 and 50% glycerol