Learn More
Enzo Life Sciences Caspase-3 polyclonal antibody (100µg)
SDP
Supplier:  Enzo Life Sciences ALX210806C100
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to aa 163-175 of human caspase-3. UniProt ID: P42574. Species reactivity: Human, Mouse, Rat. Applications: WB. Recommended Dilutions: Western Blot (0.5-4µg/ml) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Epitope-affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 1% BSA, 0.02% thimerosal and 50% glycerol. Handling: Avoid freeze/thaw cycles. After opening, prepare aliquots and store at -80°C.
Catalog No. 50-200-8162