Learn More
Enzo Life Sciences Caspase-3 polyclonal antibody (200µl)
SDP
Supplier:  Enzo Life Sciences ADIAAS103F
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to a portion of human caspase-3. The sequence differs from rat caspase-3 by 1 aa and from mouse by 2 aa. UniProt ID: P42574. Source: Whole rabbit serum. Species reactivity: Human, Mouse, Rat, Xenopus, Sheep, Rabbit, Porcine, Monkey, Hamster, Guinea pig, Dog, Chicken, Bovine. Applications: WB. Recommended Dilutions: Western Blot (ECL, 1:5,000) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Formulation: Liquid. Antiserum. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-7170