Learn More
Enzo Life Sciences Caspase-3 polyclonal antibody (50µg)
SDP
Supplier:  Enzo Life Sciences ADIAAP113D
Encompass_Preferred
Host: Rabbit. Immunogen: Recombinant human Caspase-3 protein (full length). Source: Purified from rabbit serum. Species reactivity: Human, Mouse, Rat, Yeast, Rabbit, Porcine, Monkey, Hamster, Guinea pig, Dog, Chicken, Bovine. Applications: IHC (PS), WB. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-7150