Learn More
CPC Scientific MCA-Asp-Glu-Val-Asp-Ala-Pro-Lys(DNP)-OH 1MG
SDP
Supplier:  CPC Scientific CASP059B
Encompass
Cleavage of this substrate at position P1 Asp results in continuous fluorescent assay monitored at emission wavelength of 393 nm. Substrate induces very little caspase-1 activity.ONE-LETTER SEQUENCE: Mca-DEVDAP-Lys(Dnp)MOLECULAR FORMULA: C50H62N10O22MOLECULAR WEIGHT:1155.1STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [189696-20-6], RESEARCH AREA: ApoptosisREFERENCES: M. Enari et al., Nature, 380, 723 (1996)
Catalog No. 50-210-9024