Learn More
CPC Scientific Z-Asp-Glu-Val-Asp-AFC 5MG
SDP
Catalog No. 502109030
Encompass
A selective inhibitor of CPP32-like caspases.ONE-LETTER SEQUENCE: Z-DEVD-AFCMOLECULAR FORMULA: C36H38F3N5O14MOLECULAR WEIGHT:821.71STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [1135416-11-3], RESEARCH AREA: ApoptosisREFERENCES: S. H. Inayat-Hussain et al., Hepatology, 25, 1516 (1997)
Catalog No. 50-210-9030 Supplier CPC Scientific Supplier No. CASP062B
Add to Cart
Add to Cart
A selective inhibitor of CPP32-like caspases.ONE-LETTER SEQUENCE: Z-DEVD-AFCMOLECULAR FORMULA: C36H38F3N5O14MOLECULAR WEIGHT:821.71STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [1135416-11-3], RESEARCH AREA: ApoptosisREFERENCES: S. H. Inayat-Hussain et al., Hepatology, 25, 1516 (1997)