Learn More
CPC Scientific Ac-Asp-Asn-Leu-Asp-MCA 1MG
SDP
Catalog No. 502109076
Encompass
ONE-LETTER SEQUENCE: Ac-DNLD-MCAMOLECULAR FORMULA: C30H38N6O12MOLECULAR WEIGHT:674.66STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [958001-92-8], RESEARCH AREA: Apoptosis
Catalog No. 50-210-9076 Supplier CPC Scientific Supplier No. CASP083A
ONE-LETTER SEQUENCE: Ac-DNLD-MCAMOLECULAR FORMULA: C30H38N6O12MOLECULAR WEIGHT:674.66STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [958001-92-8], RESEARCH AREA: Apoptosis