Learn More
Cayman Chemical Caspase-3/7 Inhibitor I, Purity: ≥98%, CAS: 1110670-49-9, 5 mg
SDP
Catalog No. 502969878
Encompass_Preferred
A potent, reversible, and highly selective isatin sulfonamide-based inhibitor of caspase-3 (Ki(app) = 60 nM; IC50 = 120 nM) and caspase-7 (Ki(app) = 170 nM); inhibits apoptosis in camptothecin treated Jurkat cells (IC50 = ~50 µM) and in chondrocytes (44% inhibition at 10 µM and 98% inhibition at 50 µM)
Catalog No. 50-296-9878 Supplier Cayman Chemical Supplier No. 144645 MG
A potent, reversible, and highly selective isatin sulfonamide-based inhibitor of caspase-3 (Ki(app) = 60 nM; IC50 = 120 nM) and caspase-7 (Ki(app) = 170 nM); inhibits apoptosis in camptothecin treated Jurkat cells (IC50 = ~50 µM) and in chondrocytes (44% inhibition at 10 µM and 98% inhibition at 50 µM)